Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEGF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001409.3 | NP_001400.3 | 37 | 5276 | ||
XM_006710406.3 | XP_006710469.1 | 34 | 4911 | |||
XM_011540885.1 | XP_011539187.1 | 38 | 5581 | |||
XM_011540886.1 | XP_011539188.1 | 37 | 5452 | |||
XM_011540887.2 | XP_011539189.1 | 33 | 4789 | |||
XM_017000533.1 | XP_016856022.1 | 37 | 5449 | |||
GenBank mRNA | AB011539.2 | - | 30 | 4316 | ||
AB231860.1 | - | 37 | 5277 | |||
AF086226.1 | - | 1 | 402 |