Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GCN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006836.1 | NP_006827.1 | 11 | 1041 | ||
GenBank mRNA | AK297271.1 | - | 11 | 1031 | ||
BC153881.1 | - | 11 | 1041 | 8860118 | ||
BQ929459.1 | - | 2 | 141 | |||
D86973.2 | - | 11 | 1041 | |||
U88837.1 | - | 10 | 1020 |