Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GCN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006836.1 | NP_006827.1 | 42 | 5427 | ||
GenBank mRNA | BC153881.1 | - | 42 | 5427 | 8860118 | |
D86973.2 | - | 42 | 5427 | |||
U77700.1 | - | 20 | 2892 | |||
U88836.1 | - | 3 | 385 |