Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other EEF1A1 SIRNA Assays ›
Gene Symbol:
Eef1a1
Gene Name:
eukaryotic translation elongation factor 1 alpha 1
Gene Aliases:
-
Chromosome Location:
Chr.9: 78478453 - 78481724 on Build GRCm38
UniGene ID:
Mm.138471
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
221288
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Eef1a1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_010106.2
NP_034236.2
4
680
GenBank mRNA
AK003188.1
-
4
626
AK011771.1
-
4
628
AK032914.1
-
4
626
AK050635.1
-
4
627
AK076696.1
-
4
628
AK081725.1
-
4
628
AK083361.1
-
4
628
AK088041.1
-
4
629
AK133725.1
-
4
628
AK135448.1
-
4
629
AK137223.1
-
4
628
AK141243.1
-
4
630
AK150798.1
-
4
628
AK151479.1
-
4
629
AK151773.1
-
4
628
AK152810.1
-
4
628
AK152993.1
-
4
629
AK153387.1
-
4
628
AK153402.1
-
4
628
AK153851.1
-
4
628
AK162049.1
-
4
628
AK162313.1
-
4
627
AK162440.1
-
4
632
AK163799.1
-
4
628
AK164113.1
-
4
630
AK164671.1
-
4
628
AK165938.1
-
4
628
AK166021.1
-
4
628
AK166033.1
-
4
628
AK166058.1
-
4
628
AK166089.1
-
4
628
AK166615.1
-
4
628
AK166807.1
-
4
628
AK167843.1
-
4
628
AK167873.1
-
4
627
AK168424.1
-
4
628
AK168467.1
-
4
634
AK187140.1
-
1
161
AK188863.1
-
2
322
AK206299.1
-
1
161
AK209036.1
-
1
159
AK210348.1
-
1
162
AK213893.1
-
1
161
BC003969.1
-
3
598
BC004005.1
-
3
589
3492770
BC004067.1
-
3
617
3590965
BC005660.1
-
3
618
3588966
BC018223.1
-
4
616
4012653
BC018485.1
-
4
630
3491827
BC040689.1
-
1
120
BC050124.1
-
2173
BC068258.1
-
2425
BC083069.1
-
4
616
6310448
BC092053.1
-
3
606
4008574
BC092276.1
-
3
611
3490007
BC092292.1
-
3
572
BC095965.1
-
3
606
6310404
BC108391.1
-
4
613
30242277
M22432.1
-
4
619
X13661.1
-
3
599
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_175838, BC111707, X61043, X63561, BC128723, and others.
Panther Classification:
Molecular Function -
translation factor
Gene Ontology Categories:
Function(s)
Process(es)
translation
translational elongation
protein homooligomerization
cellular response to epidermal growth factor stimulus
tRNA binding
nucleotide binding
mRNA binding
translation elongation factor activity
GTPase activity
protein binding
GTP binding
protein kinase binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-hnnbr:80/10.239.164.27:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline