Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Suclg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011507.3 | NP_035637.2 | 9 | 1115 | ||
GenBank mRNA | AF058956.1 | - | 8 | 957 | ||
AK045176.1 | - | 9 | 1068 | |||
AK156263.1 | - | 9 | 1071 | |||
AK167136.1 | - | 9 | 1070 | |||
AK167339.1 | - | 9 | 1354 | |||
AK172561.1 | - | 9 | 1070 | |||
AK172633.1 | - | 9 | 1070 | |||
BC030947.1 | - | 1 | 92 | |||
BC043312.1 | - | 7 | 810 | |||
BC054425.1 | - | 8 | 868 | |||
BC080781.1 | - | 9 | 1070 | 30470296 | ||
EU234009.1 | - | 9 | 1022 |