Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAMTA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015215.3 | NP_056030.1 | 21 | 5130 | ||
XM_011541083.2 | XP_011539385.1 | 22 | 5015 | |||
XM_011541084.2 | XP_011539386.1 | 22 | 5015 | |||
XM_011541085.2 | XP_011539387.1 | 23 | 5087 | |||
XM_011541086.2 | XP_011539388.1 | 21 | 4994 | |||
XM_011541087.2 | XP_011539389.1 | 21 | 4943 | |||
XM_011541088.2 | XP_011539390.1 | 21 | 4945 | |||
XM_011541089.2 | XP_011539391.1 | 22 | 4676 | |||
XM_011541090.2 | XP_011539392.1 | 22 | 4676 | |||
XM_017000773.1 | XP_016856262.1 | 23 | 5767 | |||
XM_017000774.1 | XP_016856263.1 | 22 | 5015 | |||
XM_017000775.1 | XP_016856264.1 | 20 | 4924 | |||
XM_017000776.1 | XP_016856265.1 | 24 | 5147 | |||
XM_017000777.1 | XP_016856266.1 | 21 | 4655 | |||
XM_017000778.1 | XP_016856267.1 | 21 | 4655 | |||
XM_017000779.1 | XP_016856268.1 | 21 | 4606 | |||
GenBank mRNA | AB020640.3 | - | 21 | 5130 | ||
AF111804.1 | - | 5 | 620 | |||
AK302088.1 | - | 14 | 2124 | |||
AK309727.1 | - | 11 | 2137 | |||
AY349360.1 | - | 5 | 615 | |||
BC051341.1 | - | 10 | 1814 | |||
BC151835.1 | - | 21 | 5130 | 8860115 |