Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAMTA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015215.3 | NP_056030.1 | 23 | 8041 | ||
XM_011541083.2 | XP_011539385.1 | 23 | 7895 | |||
XM_011541084.2 | XP_011539386.1 | 24 | 7926 | |||
XM_011541086.2 | XP_011539388.1 | 22 | 7874 | |||
XM_011541089.2 | XP_011539391.1 | 23 | 7556 | |||
XM_017000775.1 | XP_016856264.1 | 22 | 7835 | |||
XM_017000777.1 | XP_016856266.1 | 22 | 7535 | |||
XM_017000778.1 | XP_016856267.1 | 23 | 7566 | |||
GenBank mRNA | BC038183.2 | - | 1 | 2838 |