Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WSCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015253.1 | NP_056068.1 | 9 | 4604 | ||
XM_005256572.1 | XP_005256629.1 | 9 | 4692 | |||
XM_005256573.1 | XP_005256630.1 | 9 | 4667 | |||
GenBank mRNA | AB011095.1 | - | 8 | 3957 |