Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCTD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015353.2 | NP_056168.1 | 6 | 1483 | ||
NR_110834.1 | 7 | 1364 | ||||
NR_110835.1 | 8 | 1446 | ||||
GenBank mRNA | BC033329.1 | - | 7 | 1364 | ||
BC080561.1 | - | 1 | 431 | |||
D79998.1 | - | 6 | 1422 |