Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCTD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015353.2 | NP_056168.1 | 5 | 719 | ||
NR_110834.1 | 6 | 600 | ||||
NR_110835.1 | 7 | 682 | ||||
GenBank mRNA | AK307083.1 | - | 7 | 637 | ||
AK310690.1 | - | 2 | 499 | |||
BC033329.1 | - | 6 | 600 | |||
D79998.1 | - | 5 | 658 |