Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021834.1 | NP_068606.1 | 3 | 185 | ||
GenBank mRNA | AJ011299.1 | - | 3 | 185 |