Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024129.1 | NP_077043.1 | 6 | 762 | ||
XM_006241285.1 | XP_006241347.1 | 6 | 947 | |||
GenBank mRNA | BC083750.1 | - | 6 | 762 | 7192499 | |
FQ214818.1 | - | 4 | 421 | |||
FQ219808.1 | - | 6 | 795 | |||
FQ220209.1 | - | 6 | 828 | |||
FQ220915.1 | - | 2 | 115 | |||
FQ222189.1 | - | 6 | 831 | |||
FQ223165.1 | - | 2 | 202 | |||
FQ223184.1 | - | 6 | 836 | |||
FQ223539.1 | - | 1 | 54 | |||
FQ228702.1 | - | 6 | 786 | |||
FQ229041.1 | - | 6 | 850 | |||
FQ229786.1 | - | 6 | 831 | |||
X59859.1 | - | 5 | 682 | |||
Z12298.1 | - | 6 | 891 |