Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025350.3 | NP_079626.2 | 8, 9 | 1235 | ||
GenBank mRNA | AK003088.1 | - | 8, 9 | 1019 | ||
AK007330.1 | - | 2, 3 | 304 | |||
BC052661.1 | - | 8, 9 | 1249 | 30050094 |