Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pir | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301402.1 | NP_001288331.1 | 2, 3 | 333 | ||
NM_027153.3 | NP_081429.1 | 3, 4 | 462 | |||
GenBank mRNA | AK009757.1 | - | 3, 4 | 405 | ||
AK082922.1 | - | 3, 4 | 347 | |||
AK090023.1 | - | 3, 4 | 319 | |||
AK134541.1 | - | 3, 4 | 463 | |||
AK138314.1 | - | 3, 4 | 458 | |||
AK151301.1 | - | 3, 4 | 365 | |||
BC024062.1 | - | 3, 4 | 368 | 5067536 | ||
BY718294.1 | - | 3, 4 | 463 | |||
BY720928.1 | - | 3, 4 | 463 |