Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pir | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027153.3 | NP_081429.1 | 2, 3 | 368 | ||
GenBank mRNA | AK009757.1 | - | 2, 3 | 311 | ||
AK082922.1 | - | 2, 3 | 253 | |||
AK090023.1 | - | 2, 3 | 225 | |||
AK134541.1 | - | 2, 3 | 369 | |||
AK138314.1 | - | 2, 3 | 364 | |||
AK151301.1 | - | 2, 3 | 271 | |||
BC024062.1 | - | 2, 3 | 274 | 5067536 | ||
BY718294.1 | - | 2, 3 | 369 | |||
BY720928.1 | - | 2, 3 | 369 |