Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 0610010F05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027860.2 | NP_082136.2 | 4 | 575 | ||
GenBank mRNA | AK029538.1 | - | 4 | 494 | ||
AK053838.1 | - | 4 | 576 | |||
AK129454.1 | - | 4 | 542 | |||
BC055049.1 | - | 4 | 542 | |||
BC065122.1 | - | 5 | 582 | |||
BC072622.1 | - | 4 | 565 | 6821718 | ||
BC079876.1 | - | 4 | 561 | 6848269 | ||
BY712751.1 | - | 4 | 577 |