Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Utp14a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028276.1 | NP_082552.1 | 12 | 1637 | ||
GenBank mRNA | AK134784.1 | - | 12 | 1662 | ||
AK172924.1 | - | 12 | 1637 | |||
AY316163.1 | - | 12 | 1660 | |||
BC082590.1 | - | 12 | 1663 | 30625504 |