Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Top1mt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028404.2 | NP_082680.1 | 13 | 1590 | ||
GenBank mRNA | AF362952.1 | - | 13 | 1591 | ||
AK013665.1 | - | 7 | 725 | |||
AK147119.1 | - | 13 | 1593 | |||
BC138582.1 | - | 13 | 1582 | 8861602 | ||
BC138583.1 | - | 13 | 1582 | 8861603 | ||
BC145336.1 | - | 13 | 1579 | 9053892 |