Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA0753 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014804.2 | NP_055619.2 | 3 | 804 | ||
XM_006721612.2 | XP_006721675.1 | 1 | 228 | |||
XM_011524090.2 | XP_011522392.1 | 3 | 804 | |||
XM_011524091.2 | XP_011522393.1 | 3 | 2189 | |||
XM_011524095.2 | XP_011522397.1 | 3 | 536 | |||
XM_011524096.1 | XP_011522398.1 | 3 | 804 | |||
XM_011524098.2 | XP_011522400.1 | 1 | 98 | |||
XM_017025455.1 | XP_016880944.1 | 3 | 747 | |||
XM_017025456.1 | XP_016880945.1 | 1 | 228 | |||
XM_017025457.1 | XP_016880946.1 | 1 | 228 | |||
GenBank mRNA | AB018296.1 | - | 3 | 549 | ||
AK292876.1 | - | 3 | 566 | |||
AK296971.1 | - | 2 | 427 | |||
BC017566.1 | - | 3 | 563 | |||
BC113016.1 | - | 2 | 519 | 40036441 | ||
BC113017.1 | - | 2 | 519 | 40036446 |