Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZMIZ2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300959.1 | NP_001287888.1 | 18 | 4905 | ||
NM_031449.3 | NP_113637.3 | 19 | 5074 | |||
NM_174929.2 | NP_777589.2 | 17 | 4935 | |||
XM_005249866.2 | XP_005249923.1 | 19 | 5125 | |||
XM_005249867.4 | XP_005249924.1 | 19 | 5127 | |||
XM_005249868.4 | XP_005249925.1 | 19 | 6145 | |||
XM_005249869.2 | XP_005249926.1 | 19 | 5098 | |||
XM_005249870.2 | XP_005249927.1 | 18 | 5047 | |||
XM_005249871.2 | XP_005249928.1 | 19 | 5029 | |||
XM_005249872.2 | XP_005249929.1 | 18 | 5020 | |||
XM_005249873.2 | XP_005249930.1 | 19 | 5002 | |||
XM_011515566.1 | XP_011513868.1 | 13 | 4265 | |||
XM_017012674.1 | XP_016868163.1 | 19 | 5206 | |||
GenBank mRNA | AK057378.1 | - | 1 | 1939 | ||
AK090415.1 | - | 19 | 5074 | |||
AK128359.1 | - | 1 | 2956 |