Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Idh3g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031551.1 | NP_113739.1 | 10 | 862 | ||
XM_006229551.1 | XP_006229613.1 | 8 | 1284 | |||
XM_006229552.2 | XP_006229614.1 | 9 | 867 | |||
XM_006229553.2 | XP_006229615.1 | 10 | 887 | |||
XM_006229554.1 | XP_006229616.1 | 9 | 814 | |||
GenBank mRNA | BC083688.1 | - | 10 | 862 | 7189278 | |
FQ213717.1 | - | 10 | 741 | |||
FQ214482.1 | - | 10 | 829 | |||
FQ214526.1 | - | 10 | 828 | |||
FQ214664.1 | - | 10 | 803 | |||
FQ214718.1 | - | 10 | 879 | |||
FQ214896.1 | - | 10 | 813 | |||
FQ216691.1 | - | 10 | 818 | |||
FQ226989.1 | - | 9 | 739 | |||
X74125.1 | - | 10 | 788 |