Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Idh3g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031551.1 | NP_113739.1 | 11 | 999 | ||
XM_006229551.1 | XP_006229613.1 | 9 | 1421 | |||
XM_006229552.2 | XP_006229614.1 | 10 | 1004 | |||
XM_006229553.2 | XP_006229615.1 | 11 | 1024 | |||
XM_006229554.1 | XP_006229616.1 | 10 | 951 | |||
GenBank mRNA | BC083688.1 | - | 11 | 999 | 7189278 | |
FQ213717.1 | - | 11 | 878 | |||
FQ214482.1 | - | 11 | 966 | |||
FQ214526.1 | - | 11 | 965 | |||
FQ214664.1 | - | 11 | 940 | |||
FQ214718.1 | - | 11 | 1016 | |||
FQ214896.1 | - | 11 | 950 | |||
FQ216691.1 | - | 11 | 955 | |||
FQ217943.1 | - | 1 | 26 | |||
FQ226989.1 | - | 10 | 876 | |||
X74125.1 | - | 11 | 925 |