Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pon1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032077.1 | NP_114466.1 | 5 | 467 | ||
GenBank mRNA | BC091403.1 | - | 5 | 508 | 7324332 | |
CO570909.1 | - | 5 | 467 | |||
FQ209439.1 | - | 5 | 485 | |||
FQ209632.1 | - | 2 | 163 | |||
FQ211027.1 | - | 5 | 478 | |||
FQ211041.1 | - | 5 | 521 | |||
FQ218360.1 | - | 5 | 482 | |||
FQ218888.1 | - | 5 | 478 | |||
FQ219478.1 | - | 5 | 470 | |||
FQ219506.1 | - | 5 | 478 | |||
FQ219654.1 | - | 5 | 470 | |||
FQ219750.1 | - | 5 | 472 | |||
FQ219776.1 | - | 5 | 478 | |||
FQ223824.1 | - | 4 | 378 | |||
U94856.1 | - | 5 | 425 |