Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C2CD2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290474.1 | NP_001277403.1 | 5 | 1085 | ||
NM_014807.4 | NP_055622.3 | 5 | 1085 | |||
XM_006718948.3 | XP_006719011.1 | 4 | 1263 | |||
XM_011543076.2 | XP_011541378.1 | 5 | 1581 | |||
XM_017018625.1 | XP_016874114.1 | 4 | 1263 | |||
XM_017018626.1 | XP_016874115.1 | 5 | 1374 | |||
XM_017018627.1 | XP_016874116.1 | 5 | 1374 | |||
XM_017018628.1 | XP_016874117.1 | 5 | 510 | |||
GenBank mRNA | AB006623.2 | - | 5 | 550 | ||
AB094094.1 | - | 5 | 1053 | |||
AK025223.1 | - | 5 | 1053 | |||
BC022219.1 | - | 5 | 951 | 3905001 | ||
BX537572.1 | - | 2 | 386 |