Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYO1G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033054.2 | NP_149043.2 | 9 | 1352 | ||
XM_017012503.1 | XP_016867992.1 | 8 | 1111 | |||
XM_017012504.1 | XP_016867993.1 | 9 | 1347 | |||
GenBank mRNA | AB290179.1 | - | 9 | 1312 | ||
AF380932.1 | - | 8 | 1112 | |||
AF380933.1 | - | 8 | 1112 | |||
AK074050.1 | - | 8 | 4195 | |||
AK074135.1 | - | 5 | 632 | |||
AK097912.1 | - | 9 | 1343 | |||
AK131089.1 | - | 4 | 3469 | |||
AL832302.1 | - | 8 | 1107 | |||
BC113544.1 | - | 9 | 1299 | 8322596 | ||
BC117241.1 | - | 9 | 1299 | 40125792 | ||
BI910371.1 | - | 1 | 50 |