Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Setbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053099.2 | NP_444329.2 | 4 | 961 | ||
GenBank mRNA | AK129143.1 | - | 2 | 131 | ||
BC046423.1 | - | 3 | 251 | |||
BC080865.1 | - | 3 | 516 | 30531988 |