Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Setbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053099.2 | NP_444329.2 | 4 | 2335 | ||
GenBank mRNA | AK129143.1 | - | 2 | 1505 | ||
BC046423.1 | - | 3 | 1625 | |||
BC080865.1 | - | 3 | 1890 | 30531988 |