Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF418 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317027.1 | NP_001303956.1 | 7 | 3544 | ||
NM_001317028.1 | NP_001303957.1 | 7 | 3258 | |||
NM_001317029.1 | NP_001303958.1 | 7 | 3206 | |||
NM_001317030.1 | NP_001303959.1 | 4 | 3111 | |||
NM_133460.2 | NP_597717.1 | 6 | 3324 | |||
XM_017026304.1 | XP_016881793.1 | 6 | 3463 | |||
XM_017026307.1 | XP_016881796.1 | 5 | 3256 | |||
XM_017026308.1 | XP_016881797.1 | 4 | 3158 | |||
XM_017026309.1 | XP_016881798.1 | 5 | 3225 | |||
XM_017026310.1 | XP_016881799.1 | 7 | 3276 | |||
XM_017026311.1 | XP_016881800.1 | 6 | 3178 | |||
XM_017026312.1 | XP_016881801.1 | 5 | 3226 | |||
XM_017026313.1 | XP_016881802.1 | 7 | 4299 | |||
XM_017026314.1 | XP_016881803.1 | 6 | 3186 | |||
GenBank mRNA | AB075836.1 | - | 5 | 3158 | ||
AY695825.1 | - | 6 | 3309 |