Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF418 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317027.1 | NP_001303956.1 | 7 | 3624 | ||
NM_001317028.1 | NP_001303957.1 | 7 | 3338 | |||
NM_001317029.1 | NP_001303958.1 | 7 | 3286 | |||
NM_001317030.1 | NP_001303959.1 | 4 | 3191 | |||
NM_133460.2 | NP_597717.1 | 6 | 3404 | |||
XM_017026304.1 | XP_016881793.1 | 6 | 3543 | |||
XM_017026305.1 | XP_016881794.1 | 8 | 3434 | |||
XM_017026306.1 | XP_016881795.1 | 8 | 3431 | |||
XM_017026307.1 | XP_016881796.1 | 5 | 3336 | |||
XM_017026308.1 | XP_016881797.1 | 4 | 3238 | |||
XM_017026309.1 | XP_016881798.1 | 5 | 3305 | |||
XM_017026310.1 | XP_016881799.1 | 7 | 3356 | |||
XM_017026311.1 | XP_016881800.1 | 6 | 3258 | |||
XM_017026312.1 | XP_016881801.1 | 5 | 3306 | |||
XM_017026313.1 | XP_016881802.1 | 7 | 4379 | |||
XM_017026314.1 | XP_016881803.1 | 6 | 3266 | |||
GenBank mRNA | AB075836.1 | - | 5 | 3238 | ||
AY695825.1 | - | 6 | 3389 |