Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Antxr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133738.1 | NP_598499.1 | 17 | 2774 | ||
GenBank mRNA | AK009839.1 | - | 1 | 912 | ||
AK166124.1 | - | 2546 | ||||
AK170655.1 | - | 17 | 2412 | |||
AK171292.1 | - | 17 | 2448 | |||
BC003908.1 | - | 8 | 1564 | |||
BC076595.1 | - | 17 | 2774 | 30649897 |