Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Icmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133788.2 | NP_598549.1 | 5 | 2544 | ||
GenBank mRNA | AF209926.1 | - | 5 | 2326 | ||
AK154410.1 | - | 6 | 2567 | |||
BC057132.1 | - | 6 | 2563 | |||
BC110695.1 | - | 5 | 2531 | 6842587 |