Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pacs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153129.2 | NP_694769.2 | 4 | 785 | ||
GenBank mRNA | AK164770.1 | - | 5 | 771 | ||
AK171969.1 | - | 4 | 768 | |||
BC062886.1 | - | 4 | 785 | 5706409 |