Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C17orf49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142798.2 | NP_001136270.1 | 3 | 306 | ||
NM_174893.3 | NP_777553.1 | 3 | 306 | |||
GenBank mRNA | AK055800.1 | - | 2 | 906 | ||
AK295795.1 | - | 3 | 261 | |||
BC040036.1 | - | 3 | 238 | 5406966 | ||
BM011108.1 | - | 3 | 269 | |||
BU596784.1 | - | 3 | 309 | |||
CB119197.1 | - | 3 | 289 |
Gene Symbol: RNASEK-C17orf49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037717.1 | 5 | 1316 | |||
GenBank mRNA | BC040148.1 | - | 5 | 1073 | 5588971 |