Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C17orf49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142798.2 | NP_001136270.1 | 1, 2 | 150 | ||
NM_001142799.2 | NP_001136271.1 | 1, 2 | 150 | |||
NM_174893.3 | NP_777553.1 | 1, 2 | 150 | |||
GenBank mRNA | AK055800.1 | - | 1, 2 | 133 | ||
AK295795.1 | - | 1, 2 | 105 | |||
BC040036.1 | - | 1, 2 | 82 | 5406966 | ||
BM011108.1 | - | 1, 2 | 113 | |||
BU596784.1 | - | 1, 2 | 153 | |||
CB119197.1 | - | 1, 2 | 133 | |||
CN280697.1 | - | 1, 2 | 118 |
Gene Symbol: RNASEK-C17orf49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037717.1 | 3, 4 | 1160 | |||
GenBank mRNA | BC040148.1 | - | 3, 4 | 917 | 5588971 |