Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C17orf49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142798.2 | NP_001136270.1 | 6 | 814 | ||
NM_001142799.2 | NP_001136271.1 | 5 | 708 | |||
NM_174893.3 | NP_777553.1 | 6 | 810 | |||
GenBank mRNA | AK055800.1 | - | 3 | 2253 | ||
AK295795.1 | - | 6 | 765 | |||
BC040036.1 | - | 6 | 742 | 5406966 | ||
BM011108.1 | - | 6 | 778 | |||
CN280697.1 | - | 5 | 676 |
Gene Symbol: RNASEK-C17orf49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037717.1 | 8 | 1820 | |||
GenBank mRNA | BC040148.1 | - | 8 | 1577 | 5588971 |