Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Intu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175515.5 | NP_780724.3 | 3716 | |||
GenBank mRNA | AK122485.1 | - | 15 | 3256 |