Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Helz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198298.1 | NP_938040.1 | 8, 9 | 649 | ||
GenBank mRNA | AB093209.1 | - | 9, 10 | 710 | ||
BC076626.1 | - | 8, 9 | 649 | 30640711 |