Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Helz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198298.1 | NP_938040.1 | 20 | 2792 | ||
GenBank mRNA | AB093209.1 | - | 21 | 2850 | ||
AK033094.1 | - | 4 | 447 | |||
AK044226.1 | - | 5 | 651 | |||
BC076626.1 | - | 20 | 2792 | 30640711 |