Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcdha2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199504.1 | NP_955798.1 | 1 | 1002 | ||
GenBank mRNA | AB113385.1 | - | 1 | 1002 | ||
AY573976.1 | - | 1 | 1002 |