Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOMM70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014820.4 | NP_055635.3 | 2 | 874 | ||
GenBank mRNA | AB018262.1 | - | 2 | 516 | ||
AK302796.1 | - | 3 | 418 | |||
BC003633.2 | - | 2 | 540 | 3605218 | ||
BC052994.1 | - | 2 | 857 | 5549394 | ||
BC065555.1 | - | 2 | 871 |