Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SV2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323031.1 | NP_001309960.1 | 2 | 625 | ||
NM_001323032.1 | NP_001309961.1 | 2 | 934 | |||
NM_001323033.1 | NP_001309962.1 | 2 | 934 | |||
NM_001323034.1 | NP_001309963.1 | 2 | 934 | |||
NM_001323037.1 | NP_001309966.1 | 3 | 841 | |||
NM_001323038.1 | NP_001309967.1 | 2 | 753 | |||
NM_001323039.1 | NP_001309968.1 | 2 | 1026 | |||
NM_014848.5 | NP_055663.1 | 3 | 1022 | |||
XM_005254998.3 | XP_005255055.1 | 2 | 687 | |||
XM_017022761.1 | XP_016878250.1 | 2 | 638 | |||
XM_017022762.1 | XP_016878251.1 | 2 | 773 | |||
GenBank mRNA | AB018278.1 | - | 1 | 295 | ||
AF052188.1 | - | 2 | 549 | |||
BC030011.2 | - | 3 | 659 | 4826206 | ||
CR457086.1 | - | 1 | 131 |