Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP2R5E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282179.1 | NP_001269108.1 | 14 | 2419 | ||
NM_001282180.1 | NP_001269109.1 | 14 | 2407 | |||
NM_001282181.1 | NP_001269110.1 | 13 | 1789 | |||
NM_001282182.1 | NP_001269111.1 | 13 | 1744 | |||
NM_006246.3 | NP_006237.1 | 14 | 2422 | |||
NR_104104.1 | 13 | 2225 | ||||
GenBank mRNA | BC034994.1 | - | 15 | 2416 | ||
L76703.1 | - | 14 | 2400 | |||
Z69029.1 | - | 14 | 2335 |