Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127660.1 | NP_001121132.1 | 3 | 600 | ||
NM_014874.3 | NP_055689.1 | 4 | 745 | |||
XM_005263543.3 | XP_005263600.1 | 5 | 758 | |||
XM_005263545.3 | XP_005263602.1 | 4 | 608 | |||
XM_005263547.3 | XP_005263604.1 | 4 | 640 | |||
XM_005263548.3 | XP_005263605.1 | 5 | 775 | |||
GenBank mRNA | AF036536.2 | - | 4 | 614 | ||
AK289828.1 | - | 3 | 545 | |||
AY028429.1 | - | 3 | 318 | |||
BC017061.1 | - | 4 | 475 | 3901235 | ||
BU178912.1 | - | 4 | 540 | |||
D86987.1 | - | 4 | 614 |