Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007231.2 | NP_001007232.2 | 2 | 664 | ||
NM_001166276.1 | NP_001159748.1 | 1 | 604 | |||
NM_001166277.1 | NP_001159749.1 | 2 | 664 | |||
NM_014882.2 | NP_055697.1 | 1 | 604 | |||
XM_005264675.2 | XP_005264732.1 | 2 | 371 | |||
XM_005264676.2 | XP_005264733.1 | 2 | 369 | |||
XM_011533206.2 | XP_011531508.1 | 2 | 412 | |||
XM_011533207.2 | XP_011531509.1 | 2 | 378 | |||
XM_011533208.2 | XP_011531510.1 | 3 | 429 | |||
XM_017005423.1 | XP_016860912.1 | 2 | 272 | |||
XM_017005424.1 | XP_016860913.1 | 2 | 272 | |||
XM_017005425.1 | XP_016860914.1 | 2 | 272 | |||
XM_017005428.1 | XP_016860917.1 | 2 | 272 | |||
GenBank mRNA | AK290396.1 | - | 2 | 342 | ||
AK297056.1 | - | 2 | 542 | |||
AK297157.1 | - | 2 | 407 | |||
AK298669.1 | - | 1 | 431 | |||
AK303591.1 | - | 3 | 499 | |||
AK310742.1 | - | 2 | 369 | |||
BC039591.1 | - | 1 | 465 | 5757311 | ||
BX443252.2 | - | 2 | 609 | |||
D29642.1 | - | 1 | 417 | |||
EU794656.1 | - | 1 | 604 |