Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1264 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000456.1 | NP_001000456.1 | 1 | 134 |
Gene Symbol: LOC100910077 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749183.3 | XP_003749231.1 | 1 | 155 |
Gene Symbol: LOC100910378 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749205.3 | XP_003749253.1 | 1 | 155 |