Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1765 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000489.1 | NP_001000489.1 | 1 | 800 |
Gene Symbol: LOC103689939 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773554.1 | XP_008771776.1 | 569 |
Set Membership: |
G-protein coupled receptor |