Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSC1A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006511.2 | NP_006502.1 | 1 | 614 | ||
GenBank mRNA | AK314753.1 | - | 1 | 614 | ||
CR999543.1 | - | 1 | 389 |
Gene Symbol: DDI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032341.4 | NP_115717.3 | 10 | 1949 | ||
GenBank mRNA | AK057374.1 | - | 8 | 1345 |