Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr859 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000585.1 | NP_001000585.1 | 1 | 605 |
Gene Symbol: LOC100910682 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750017.2 | XP_003750065.1 | 605 |