Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA0753 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014804.2 | NP_055619.2 | 5 | 1207 | ||
XM_011524090.2 | XP_011522392.1 | 5 | 1207 | |||
XM_011524091.2 | XP_011522393.1 | 5 | 2592 | |||
XM_011524095.2 | XP_011522397.1 | 5 | 939 | |||
XM_011524096.1 | XP_011522398.1 | 5 | 1207 | |||
XM_011524100.2 | XP_011522402.1 | 2 | 110 | |||
XM_017025455.1 | XP_016880944.1 | 5 | 1150 | |||
GenBank mRNA | AB018296.1 | - | 5 | 952 | ||
AK292876.1 | - | 5 | 969 | |||
BC017566.1 | - | 5 | 966 | |||
BC113016.1 | - | 4 | 922 | 40036441 | ||
BC113017.1 | - | 3 | 815 | 40036446 |