Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA0319 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168374.1 | NP_001161846.1 | 22 | 4822 | ||
NM_001168375.1 | NP_001161847.1 | 21 | 4706 | |||
NM_001168376.1 | NP_001161848.1 | 20 | 4546 | |||
NM_001168377.1 | NP_001161849.1 | 19 | 4548 | |||
NM_001252328.1 | NP_001239257.1 | 17 | 3241 | |||
NM_014809.3 | NP_055624.2 | 21 | 4731 | |||
XM_011515025.2 | XP_011513327.1 | 21 | 4488 | |||
XM_011515026.2 | XP_011513328.1 | 20 | 4449 | |||
XM_017011541.1 | XP_016867030.1 | 22 | 4802 | |||
XM_017011543.1 | XP_016867032.1 | 20 | 4350 | |||
XM_017011544.1 | XP_016867033.1 | 20 | 4423 | |||
XM_017011545.1 | XP_016867034.1 | 21 | 4444 | |||
XM_017011547.1 | XP_016867036.1 | 20 | 4358 | |||
XM_017011548.1 | XP_016867037.1 | 20 | 4431 | |||
XM_017011549.1 | XP_016867038.1 | 20 | 4388 | |||
XM_017011551.1 | XP_016867040.1 | 19 | 4235 | |||
XM_017011552.1 | XP_016867041.1 | 22 | 4406 | |||
XM_017011553.1 | XP_016867042.1 | 22 | 4248 | |||
GenBank mRNA | AB002317.1 | - | 21 | 4717 | ||
BC140821.1 | - | 22 | 4562 | 9021689 | ||
BC144628.1 | - | 19 | 4407 | 9053159 | ||
BC152460.1 | - | 21 | 4717 | 8862596 |